4-(Benzyloxy)-1-iodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59764-500MG 59764-1G 59764-5G 59764-25G
77330-676EA 252.35 USD
77330-676 77354-830 77366-896 77388-510
4-(Benzyloxy)-1-iodo-2-methylbenzene ≥97%
4-(Benzyloxy)-1-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃IO
Storage Temperature: Ambient
MDL Number: MFCD32649096
CAS Number: 1149618-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR