3-Propoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

59757-500MG 59757-1G 59757-5G 59757-25G
77330-674EA 278.1 USD
77330-674 77354-828 77366-894 77388-508
3-Propoxy-5-(trifluoromethyl)benzoic acid ≥97%
3-Propoxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34759390
CAS Number: 2755721-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR