(4-Chloro-3-ethylphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

59754-500MG 59754-1G 59754-5G 59754-25G
77330-672EA 258.53 USD
77330-672 77354-826 77366-892 77388-506
(4-Chloro-3-ethylphenyl)(morpholino)methanone ≥97%
(4-Chloro-3-ethylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD34759388
CAS Number: 2755719-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR