4-Chloro-3-ethyl-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

59753-500MG 59753-1G 59753-5G 59753-25G
77330-670EA 226.6 USD
77330-670 77354-824 77366-890 77388-504
4-Chloro-3-ethyl-N-methylbenzamide ≥97%
4-Chloro-3-ethyl-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO
MW: 197.661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759387
CAS Number: 2755724-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR