4-Chloro-3-ethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

59750-500MG 59750-1G 59750-5G 59750-25G
77330-666EA 235.87 USD
77330-666 77354-820 77366-886 77388-500
4-Chloro-3-ethylbenzamide ≥97%
4-Chloro-3-ethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD19439989
CAS Number: 1369828-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR