Ethyl 4-chloro-3-ethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

59748-500MG 59748-1G 59748-5G 59748-25G
77330-664EA 175.1 USD
77330-664 77346-488 77366-884 77388-498
Ethyl 4-chloro-3-ethylbenzoate ≥97%
Ethyl 4-chloro-3-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂
MW: 212.673 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19439985
CAS Number: 1379358-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR