(3-Bromo-2-fluoro-6-propoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

59743-500MG 59743-1G 59743-5G 59743-25G
77330-662EA 126.69 USD
77330-662 77344-168 77366-882 77378-948
(3-Bromo-2-fluoro-6-propoxyphenyl)(methyl)sulfane ≥95%
(3-Bromo-2-fluoro-6-propoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFOS
MW: 279.169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759383
CAS Number: 2764733-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR