(3-Bromo-2-fluoro-6-propoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59742-500MG 59742-1G 59742-5G 59742-25G
77330-660EA 186.43 USD
77330-660 77354-818 77366-880 77388-496
(3-Bromo-2-fluoro-6-propoxyphenyl)methanol ≥97%
(3-Bromo-2-fluoro-6-propoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759382
CAS Number: 2764731-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR