3-Bromo-6-butoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

59735-500MG 59735-1G 59735-5G 59735-25G
77330-656EA 128.75 USD
77330-656 77344-262 77366-876 77378-944
3-Bromo-6-butoxy-2-fluorobenzaldehyde ≥97%
3-Bromo-6-butoxy-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34759377
CAS Number: 2755723-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR