Ethyl 4-butoxy-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

59728-500MG 59728-1G 59728-5G 59728-25G
77330-650EA 120.51 USD
77330-650 77343-764 77366-870 77378-938
Ethyl 4-butoxy-2-fluorobenzoate ≥97%
Ethyl 4-butoxy-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD34759372
CAS Number: 2807470-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR