1-(Benzyloxy)-4-bromo-3-fluoro-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

59725-500MG 59725-1G 59725-5G 59725-25G
77330-646EA 136.99 USD
77330-646 77344-546 77366-866 77388-494
1-(Benzyloxy)-4-bromo-3-fluoro-2-iodobenzene ≥95%
1-(Benzyloxy)-4-bromo-3-fluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrFIO
Storage Temperature: Ambient
MDL Number: MFCD34759370
CAS Number: 2131213-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR