(6-(Benzyloxy)-3-bromo-2-fluorophenyl)(methyl)sulphane ≥97%
Supplier: AOBChem USA
|
Warning
|
59724-500MG
59724-1G
59724-5G
59724-25G
77330-644EA
145.23
USD
77330-644
77345-098
77366-864
77388-492
(6-(Benzyloxy)-3-bromo-2-fluorophenyl)(methyl)sulphane ≥97%
(6-(Benzyloxy)-3-bromo-2-fluorophenyl)(methyl)sulphane
Precaución: for Research Use Only
Formula:
C₁₄H₁₂BrFOS Storage Temperature: Ambient |
MDL Number:
MFCD34759369 CAS Number: 2755724-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...