Ethyl 4-ethoxy-3-ethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

59716-500MG 59716-1G 59716-5G
77338-498EA 491.31 USD
77338-498 77354-816 77378-930
Ethyl 4-ethoxy-3-ethylbenzoate ≥97%
Ethyl 4-ethoxy-3-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759364
CAS Number: 2383232-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR