3-(3-Bromo-6-chloro-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%

Supplier: AOBChem USA

Warning

59711-500MG 59711-1G 59711-5G 59711-25G
77330-636EA 145.23 USD
77330-636 77345-096 77366-856 77388-488
3-(3-Bromo-6-chloro-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%
3-(3-Bromo-6-chloro-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrClFNO₃
MW: 340.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759359
CAS Number: 2755719-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR