3-(3-Bromo-5-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%

Supplier: AOBChem USA

Warning

59708-1G 59708-5G 59708-25G
77341-940EA 108.15 USD
77341-940 77366-854 77378-928
3-(3-Bromo-5-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%
3-(3-Bromo-5-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrFNO₃
MW: 306.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759356
CAS Number: 2755723-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR