3-(2-Bromo-6-fluoro-4-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59700-500MG 59700-1G 59700-5G 59700-25G
77330-632EA 140.08 USD
77330-632 77344-722 77366-848 77388-484
3-(2-Bromo-6-fluoro-4-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(2-Bromo-6-fluoro-4-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrFNO₃
MW: 320.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759348
CAS Number: 2755718-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR