Ethyl 3-(2,4-bis(trifluoromethyl)phenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

59695-1G 59695-5G 59695-25G
77340-760EA 60.77 USD
77340-760 77366-844 77378-922
Ethyl 3-(2,4-bis(trifluoromethyl)phenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(2,4-bis(trifluoromethyl)phenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₂F₆O₃
Storage Temperature: Ambient
MDL Number: MFCD34759344
CAS Number: 2901083-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR