(3-Bromo-6-ethoxy-2-fluorophenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

59690-500MG 59690-1G 59690-5G 59690-25G
77330-626EA 162.74 USD
77330-626 77346-094 77366-838 77388-480
(3-Bromo-6-ethoxy-2-fluorophenyl)(methyl)sulphane ≥97%
(3-Bromo-6-ethoxy-2-fluorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₉H₁₀BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34759340
CAS Number: 2708591-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR