3-Bromo-6-ethoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

59685-500MG 59685-1G 59685-5G 59685-25G
77330-624EA 100.94 USD
77330-624 77342-952 77366-836 77378-918
3-Bromo-6-ethoxy-2-fluorobenzoic acid ≥97%
3-Bromo-6-ethoxy-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34759335
CAS Number: 2386461-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR