3-(3-Fluoro-5-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59678-1G 59678-5G 59678-25G
77341-628EA 96.82 USD
77341-628 77366-830 77378-914
3-(3-Fluoro-5-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(3-Fluoro-5-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆FNO₃
MW: 241.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759329
CAS Number: 2755723-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR