3-(4-Bromo-3-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%

Supplier: AOBChem USA

Warning

59676-500MG 59676-1G 59676-5G 59676-25G
77330-618EA 94.76 USD
77330-618 77342-530 77366-826 77378-910
3-(4-Bromo-3-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%
3-(4-Bromo-3-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrF₃NO₃
MW: 356.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759327
CAS Number: 2755722-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR