3-(2-Bromo-5-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%

Supplier: AOBChem USA

Warning

59670-500MG 59670-1G 59670-5G 59670-25G
77330-612EA 99.91 USD
77330-612 77342-842 77366-820 77378-904
3-(2-Bromo-5-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%
3-(2-Bromo-5-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₃
MW: 302.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759322
CAS Number: 2755718-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR