3-(4-(Benzyloxy)-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59669-500MG 59669-1G 59669-5G 59669-25G
77330-610EA 140.08 USD
77330-610 77344-720 77366-818 77388-476
3-(4-(Benzyloxy)-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(4-(Benzyloxy)-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₉H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD34759321
CAS Number: 2755723-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR