3-(3-(Benzyloxy)-4-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
Supplier: AOBChem USA
|
Warning
|
59668-500MG
59668-1G
59668-5G
59668-25G
77330-608EA
100.94
USD
77330-608
77342-950
77366-816
77378-902
3-(3-(Benzyloxy)-4-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(3-(Benzyloxy)-4-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide
Precaución: for Research Use Only
Formula:
C₁₉H₂₃NO₄ MW: 329.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759320 CAS Number: 2807464-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...