3-(4-Chloro-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59662-500MG 59662-1G 59662-5G 59662-25G
77330-604EA 125.66 USD
77330-604 77344-092 77366-812 77378-900
3-(4-Chloro-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(4-Chloro-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClFNO₃
MW: 261.677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759316
CAS Number: 2755723-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR