3-(4-Bromo-2-ethylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%
Supplier: AOBChem USA
|
Warning
|
59659-500MG
59659-1G
59659-5G
59659-25G
77330-600EA
257.5
USD
77330-600
77354-810
77366-808
77388-470
3-(4-Bromo-2-ethylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%
3-(4-Bromo-2-ethylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BrNO₃ MW: 316.191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759313 CAS Number: 2807471-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...