1-Bromo-2-ethoxy-5-fluoro-3-(trifluoromethyl)benzene ≥95%
Supplier: AOBChem USA
|
Warning
|
59646-500MG
59646-1G
59646-5G
59646-25G
77330-596EA
97.85
USD
77330-596
77342-676
77366-804
77378-898
1-Bromo-2-ethoxy-5-fluoro-3-(trifluoromethyl)benzene ≥95%
1-Bromo-2-ethoxy-5-fluoro-3-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₉H₇BrF₄O MW: 287.049 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759301 CAS Number: 2386509-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...