1-Bromo-3-fluoro-2-methoxy-4-propoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
59627-500MG
59627-1G
59627-5G
59627-25G
77330-588EA
101.97
USD
77330-588
77343-004
77366-798
77378-894
1-Bromo-3-fluoro-2-methoxy-4-propoxybenzene ≥97%
1-Bromo-3-fluoro-2-methoxy-4-propoxybenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₂BrFO₂ MW: 263.103 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759287 CAS Number: 2807446-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...