3-Formyl-2-isopropoxy-5-isopropylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
59597-500MG
59597-1G
59597-5G
77340-008EA
712.76
USD
77340-008
77354-798
77378-890
3-Formyl-2-isopropoxy-5-isopropylbenzoic acid ≥97%
3-Formyl-2-isopropoxy-5-isopropylbenzoic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₈O₄ MW: 250.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759262 CAS Number: 2807439-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...