Methyl 3-bromo-5-isopropyl-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

59592-500MG 59592-1G 59592-5G
77337-168EA 408.91 USD
77337-168 77354-794 77378-886
Methyl 3-bromo-5-isopropyl-2-methoxybenzoate ≥95%
Methyl 3-bromo-5-isopropyl-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₃
MW: 287.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17167557
CAS Number: 1245532-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR