Ethyl 3-bromo-5-isopropyl-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

59591-500MG 59591-1G 59591-5G 59591-25G
77330-582EA 264.71 USD
77330-582 77354-792 77366-792 77388-460
Ethyl 3-bromo-5-isopropyl-2-methoxybenzoate ≥95%
Ethyl 3-bromo-5-isopropyl-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇BrO₃
MW: 301.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17167621
CAS Number: 1245529-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR