Ethyl 3-ethyl-4-isopropoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

59587-500MG 59587-1G 59587-5G
77338-356EA 481.01 USD
77338-356 77354-790 77378-884
Ethyl 3-ethyl-4-isopropoxybenzoate ≥97%
Ethyl 3-ethyl-4-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₃
MW: 236.307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759254
CAS Number: 2807463-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR