4-Bromo-2-ethyl-1-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

59576-500MG 59576-1G 59576-5G
77330-578EA 337.84 USD
77330-578 77354-780 77378-876
4-Bromo-2-ethyl-1-isopropoxybenzene ≥97%
4-Bromo-2-ethyl-1-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356525
CAS Number: 749932-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR