2-Fluoro-3-methyl-6-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

59575-500MG 59575-1G 59575-5G
77336-836EA 394.49 USD
77336-836 77354-778 77378-874
2-Fluoro-3-methyl-6-(trifluoromethyl)phenol ≥97%
2-Fluoro-3-methyl-6-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MW: 194.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759246
CAS Number: 2386174-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR