2-Fluoro-3-iodo-6-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

59573-500MG 59573-1G 59573-5G
77340-338EA 908.46 USD
77340-338 77354-776 77378-872
2-Fluoro-3-iodo-6-(trifluoromethyl)phenol ≥97%
2-Fluoro-3-iodo-6-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃F₄IO
Storage Temperature: Ambient
MDL Number: MFCD34759244
CAS Number: 2384099-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR