Methyl 4-bromo-2-ethyl-6-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

59557-500MG 59557-1G 59557-5G
77336-220EA 383.16 USD
77336-220 77354-768 77378-864
Methyl 4-bromo-2-ethyl-6-methylbenzoate ≥95%
Methyl 4-bromo-2-ethyl-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23707511
CAS Number: 1427409-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR