5-(3-Bromo-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59554-500MG 59554-1G 59554-5G 59554-25G
77330-576EA 134.93 USD
77330-576 77344-472 77366-788 77388-456
5-(3-Bromo-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(3-Bromo-5-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂S
MW: 323.132 g/mol
Storage Temperature: Ambient
CAS Number: 2807469-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR