3-Chloro-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

59525-500MG 59525-1G 59525-5G
77340-266EA 865.2 USD
77340-266 77354-766 77378-862
3-Chloro-2-(methylthio)benzoic acid ≥97%
3-Chloro-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD16036854
CAS Number: 147460-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR