4-Chloro-3-ethyl-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

59505-500MG 59505-1G 59505-5G
77338-460EA 489.25 USD
77338-460 77354-756 77378-854
4-Chloro-3-ethyl-5-methylbenzoic acid ≥97%
4-Chloro-3-ethyl-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759198
CAS Number: 2705738-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR