4-Chloro-3-ethyl-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

59503-500MG 59503-1G 59503-5G
77336-834EA 394.49 USD
77336-834 77354-754 77378-852
4-Chloro-3-ethyl-5-methylbenzaldehyde ≥97%
4-Chloro-3-ethyl-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD34759196
CAS Number: 2711282-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR