Ethyl 4-(benzyloxy)-2,5-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

59488-500MG 59488-1G 59488-5G 59488-25G
77330-566EA 200.85 USD
77330-566 77354-752 77366-778 77388-452
Ethyl 4-(benzyloxy)-2,5-dichlorobenzoate ≥97%
Ethyl 4-(benzyloxy)-2,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₄Cl₂O₃
MW: 325.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759193
CAS Number: 2807444-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR