Methyl 4-(benzyloxy)-2,5-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

59487-500MG 59487-1G 59487-5G 59487-25G
77330-564EA 174.07 USD
77330-564 77346-454 77366-776 77388-450
Methyl 4-(benzyloxy)-2,5-dichlorobenzoate ≥97%
Methyl 4-(benzyloxy)-2,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₂Cl₂O₃
MW: 311.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759192
CAS Number: 2807439-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR