4-(Benzyloxy)-2,5-dichlorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

59484-500MG 59484-1G 59484-5G 59484-25G
77330-562EA 128.75 USD
77330-562 77344-260 77366-774 77378-844
4-(Benzyloxy)-2,5-dichlorobenzaldehyde ≥95%
4-(Benzyloxy)-2,5-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34759189
CAS Number: 2764731-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR