Methyl 2,5-dichloro-4-(difluoromethoxy)benzoate ≥95%

Supplier: AOBChem USA

Warning

59475-500MG 59475-1G 59475-5G
77335-756EA 364.62 USD
77335-756 77354-744 77378-838
Methyl 2,5-dichloro-4-(difluoromethoxy)benzoate ≥95%
Methyl 2,5-dichloro-4-(difluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₆Cl₂F₂O₃
MW: 271.045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28785320
CAS Number: 1806297-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR