3-(5-Bromo-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59471-1G 59471-5G 59471-25G
77341-246EA 78.28 USD
77341-246 77366-770 77378-836
3-(5-Bromo-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(5-Bromo-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrFNO₃
MW: 306.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759183
CAS Number: 2764729-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR