3-(2-Bromo-4-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59470-500MG 59470-1G 59470-5G 59470-25G
77330-556EA 204.97 USD
77330-556 77354-742 77366-768 77388-444
3-(2-Bromo-4-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(2-Bromo-4-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrF₃NO₃
MW: 356.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759182
CAS Number: 2807464-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR