3-(2-Bromo-4-chlorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59469-500MG 59469-1G 59469-5G 59469-25G
77330-554EA 100.94 USD
77330-554 77342-948 77366-766 77378-834
3-(2-Bromo-4-chlorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(2-Bromo-4-chlorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrClNO₃
MW: 322.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759181
CAS Number: 2755717-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR