3-(2-Bromo-4-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
Supplier: AOBChem USA
|
Warning
|
59468-500MG
59468-1G
59468-5G
59468-25G
77330-552EA
100.94
USD
77330-552
77342-946
77366-764
77378-832
3-(2-Bromo-4-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(2-Bromo-4-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide
Precaución: for Research Use Only
Formula:
C₁₁H₁₃BrFNO₃ MW: 306.128 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759180 CAS Number: 2755720-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...