3-(5-Bromo-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59467-500MG 59467-1G 59467-5G 59467-25G
77330-550EA 155.53 USD
77330-550 77345-568 77366-762 77388-442
3-(5-Bromo-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(5-Bromo-2-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₃
MW: 302.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759179
CAS Number: 2755718-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR