3-(4-Bromo-3-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59466-500MG 59466-1G 59466-5G 59466-25G
77330-548EA 126.69 USD
77330-548 77344-162 77366-760 77378-830
3-(4-Bromo-3-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(4-Bromo-3-methylphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₃
MW: 302.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759178
CAS Number: 2755718-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR